Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human LAMP-2/CD107b Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC6228G
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC6228G 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC6228G Supplier R&D Systems Supplier No. IC6228G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

LAMP-2/CD107b Monoclonal specifically detects LAMP-2/CD107b in Human samples. It is validated for Intracellular Staining by Flow Cytometry, Immunocytochemistry.

Specifications

Antigen LAMP-2/CD107b
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 743320
Conjugate Alexa Fluor 488
Dilution Intracellular Staining by Flow Cytometry 5 uL/10^6 cells, Immunocytochemistry 8-25 ug/mL
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. P13473
Gene Alias CD107 antigen-like family member B, CD107b, CD107b antigen, LAMP2, LAMP-2, LAMPB, LGP110, LGP-96, Lysosomal Associated Membrane Protein 2, lysosomal-associated membrane protein 2, lysosome-associated membrane glycoprotein 2, Lysosome-associated membrane p
Gene Symbols LAMP2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human LAMP2/CD107b Leu29-Phe375 Accession # P13473
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3920
Test Specificity Conjugated LAMP2/CD107b antibodies are ideal for immunocytochemistry colocalization studies in lysosomes. The unconjugated antibody detects human LAMP2/CD107b in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human LAMP1 is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.