Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human LAMP Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF873
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF873 100 μg
AF873SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF873 Supplier R&D Systems Supplier No. AF873
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

LAMP Polyclonal specifically detects LAMP in Human samples. It is validated for Western Blot.

Specifications

Antigen LAMP
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Gene Accession No. AAC50569
Gene Alias FLJ34254, FLJ35396, IgLON family member 3, IGLON3FLJ54658, LAMPFLJ37216, limbic system-associated membrane protein, LSAMP
Gene Symbols LSAMP
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human LAMP Val29-Asn315 Accession # AAC50569
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4045
Test Specificity Detects human LAMP in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.