Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human LAMP Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF873
This item is not returnable.
View return policy
Description
LAMP Polyclonal specifically detects LAMP in Human samples. It is validated for Western Blot.Specifications
LAMP | |
Polyclonal | |
Biotin | |
AAC50569 | |
LSAMP | |
Mouse myeloma cell line NS0-derived recombinant human LAMP Val29-Asn315 Accession # AAC50569 | |
50 μg | |
Primary | |
Detects human LAMP in Western blots. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
FLJ34254, FLJ35396, IgLON family member 3, IGLON3FLJ54658, LAMPFLJ37216, limbic system-associated membrane protein, LSAMP | |
Goat | |
Affinity Purified | |
RUO | |
4045 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction