Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human LAP (TGF-beta 1) Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB246100
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB246100 100 μg
MAB246SP 25 μg
MAB246500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB246100 Supplier R&D Systems Supplier No. MAB246100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 8 publications

LAP (TGF-beta 1) Monoclonal specifically detects LAP (TGF-beta 1) in Human samples. It is validated for Neutralization, Immunocytochemistry.

Specifications

Antigen LAP (TGF-beta 1)
Applications Neutralization, Immunocytochemistry
Classification Monoclonal
Clone 27235
Conjugate Unconjugated
Dilution Neutralization 1-5 ug/mL, Immunocytochemistry 3-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias CED, DPD1, LAP, LAP (TGFbeta 1), TGFB, TGFB1, TGFbeta, transforming growth factor beta 1
Gene Symbols TGFB1
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human LAP (TGF-beta 1)
Purification Method Protein A or G purified from ascites
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7040
Test Specificity Detects human LAP in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.