Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human LARG Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF4737
This item is not returnable.
View return policy
Description
LARG Polyclonal specifically detects LARG in Human samples. It is validated for Western Blot.Specifications
LARG | |
Polyclonal | |
Unconjugated | |
Q9NZN5 | |
ARHGEF12 | |
E. coli-derived recombinant human LARG Ile154-Leu575 Accession # Q9NZN5 | |
100 μg | |
Primary | |
Detects endogenous human LARG in Western blots. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
ARHGC, ARHGEF12, DKFZp686O2372, KIAA0382PRO2792, LARGleukemia-associated rho guanine nucleotide exchange factor, Leukemia-associated RhoGEF, Rho guanine nucleotide exchange factor (GEF) 12, rho guanine nucleotide exchange factor 12 | |
Goat | |
Affinity Purified | |
RUO | |
23365 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction