Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Latent TGF-beta bp2/LTBP-2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3850SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3850SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3850SP Supplier R&D Systems Supplier No. AF3850SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Latent TGF-beta bp2/LTBP-2 Polyclonal specifically detects Latent TGF-beta bp2/LTBP-2 in Human samples. It is validated for Western Blot.

Specifications

Antigen Latent TGF-beta bp2/LTBP-2
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q14767
Gene Alias C14orf141, GLC3D, Latent TGFbeta bp2, latent transforming growth factor beta binding protein 2, latent-transforming growth factor beta-binding protein 2, LTBP2, LTBP-2, LTBP3chromosome 14 open reading frame 141, MSTP031
Gene Symbols LTBP2
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Latent TGF-beta bp2/LTBP-2 Gln36-Glu1821 Accession # Q14767
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4053
Test Specificity Detects human Latent TGF-beta bp2/LTBP-2 in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant mouse (rm) LTBP-2 is observed, and less than 1% cross-reactivity with recombinant human LTBP-1 and rmLTBP-4 is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.