Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Latexin Antibody, R&D Systems™

Goat Polyclonal Antibody
$164.60 - $496.43
Specifications
| Antigen | Latexin |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
Description
Latexin Polyclonal specifically detects Latexin in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| Latexin | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| ECI, ECIMUM, Endogenous carboxypeptidase inhibitor, latexin, LXN, TCI, TCIProtein MUM, Tissue carboxypeptidase inhibitor | |
| LXN | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| AAH05346 | |
| 56925 | |
| E. coli-derived recombinant human Latexin Glu2-Glu222 Accession # AAH05346 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human Latexin in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title