Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Latrophilin 2/LPHN2 Alexa Fluor™ 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB105881R1
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB105881R1 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB105881R1 Supplier R&D Systems Supplier No. FAB105881R100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Latrophilin 2/LPHN2 Monoclonal antibody specifically detects Latrophilin 2/LPHN2 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen Latrophilin 2/LPHN2
Applications Flow Cytometry
Classification Monoclonal
Clone 1030202
Conjugate Alexa Fluor 647
Dilution Flow Cytometry 0.25-1 ug/106 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. O95490
Gene Alias ADGRL2, Calcium-independent alpha-latrotoxin receptor 2, CIRL2, CIRL-2, CL2, KIAA0786, latrophilin 2, Latrophilin homolog 1, latrophilin homolog 2 (cow), latrophilin-2, LEC1, LEC1LPHH1latrophilin 1, lectomedin-1, LPHH1, LPHN2
Host Species Mouse
Immunogen Human Latrophilin 2/LPHN2 synthetic peptide, Accession # O95490
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23266
Reconstitution Reconstitution default not known
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.