Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Layilin Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $655.00
Specifications
Antigen | Layilin |
---|---|
Clone | 328002 |
Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB26371SP
![]() |
R&D Systems
MAB26371SP |
25 μg |
Each for $200.50
|
|
|||||
MAB26371
![]() |
R&D Systems
MAB26371 |
500 μg |
Each for $655.00
|
|
|||||
Description
Layilin Monoclonal specifically detects Layilin in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
Layilin | |
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FLJ30977, FLJ31092, layilin, LAYN, LYAN | |
LAYN | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
328002 | |
ELISA | |
Unconjugated | |
Mouse | |
Human | |
NP_849156 | |
143903 | |
Mouse myeloma cell line NS0-derived recombinant human Layilin Ala22-Glu220 Accession # NP_849156 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Layilin in ELISAs. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title