Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human LC3A Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB8558
This item is not returnable.
View return policy
Description
LC3A Monoclonal specifically detects LC3A in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
LC3A | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Apg8, APG8a, Apg8p3, ATG8E, Autophagy-related protein LC3 A, Autophagy-related ubiquitin-like modifier LC3 A, LC3, LC3A, MAP1 light chain 3-like protein 1, MAP1A/1B light chain 3 A, MAP1A/MAP1B LC3 A, MAP1A/MAP1B light chain 3 A, MAP1ALC3, MAP1BLC3, MAP1LC3A, Microtubule-associated protein 1 light chain 3 alpha, microtubule-associated proteins 1A/1B light chain 3, microtubule-associated proteins 1A/1B light chain 3A, MLP3A | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
84557 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Western Blot, Immunohistochemistry | |
877005 | |
Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL | |
Q9H492 | |
MAP1LC3A | |
E. coli-derived recombinant human LC3A Accession # Q9H492 | |
100 μg | |
Primary | |
Detects human LC3A in direct ELISAs. | |
Human | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction