Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human LC3A Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$200.50 - $410.00
Specifications
Antigen | LC3A |
---|---|
Clone | 2312D |
Dilution | Western Blot 1μg/mL, Immunohistochemistry 3-25μg/mL |
Applications | Immunohistochemistry, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9915-SP
![]() |
R&D Systems
MAB9915SP |
25 μg |
Each for $200.50
|
|
|||||
MAB9915-100
![]() |
R&D Systems
MAB9915100 |
100 μg |
Each for $410.00
|
|
|||||
Description
LC3A Monoclonal specifically detects LC3A in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
LC3A | |
Western Blot 1μg/mL, Immunohistochemistry 3-25μg/mL | |
Monoclonal | |
Supernatant | |
Human | |
Q9H492 | |
84557 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
2312D | |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
Apg8, APG8a, Apg8p3, ATG8E, Autophagy-related protein LC3 A, Autophagy-related ubiquitin-like modifier LC3 A, LC3, LC3A, MAP1 light chain 3-like protein 1, MAP1A/1B light chain 3 A, MAP1A/MAP1B LC3 A, MAP1A/MAP1B light chain 3 A, MAP1ALC3, MAP1BLC3, MAP1LC3A, Microtubule-associated protein 1 light chain 3 alpha, microtubule-associated proteins 1A/1B light chain 3, microtubule-associated proteins 1A/1B light chain 3A, MLP3A | |
Synthetic peptide containing human LC3A peptide Accession # Q9H492 | |
Primary | |
Reconstitute at 0.5mg/mL in sterile PBS. | |
Detects human LC3A in direct ELISA. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title