Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Lck Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC37041U100UG
This item is not returnable.
View return policy
Description
Lck Monoclonal specifically detects Lck in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
Lck | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
P06239 | |
LCK | |
E. coli-derived recombinant human Lck Met1-Val66 Accession # P06239 | |
RUO | |
3932 | |
Human | |
IgG2a |
Flow Cytometry | |
693010 | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
EC 2.7.10, EC 2.7.10.2, Hck-3, Lcktkr, Leukocyte C-terminal Src kinase, LSK, LSTRA, Lymphocyte cell-specific protein-tyrosine kinase, lymphocyte-specific protein tyrosine kinase, p56(LSTRA) protein-tyrosine kinase, p56lck, p56-LCK, pp58lck, Protein YT16, Proto-oncogene Lck, proto-oncogene tyrosine-protein kinase LCK, T cell-specific protein-tyrosine kinase, T-lymphocyte specific protein tyrosine kinase p56lck, tyrosine-protein kinase Lck, YT16 | |
Mouse | |
100 μg | |
Primary | |
Detects human Lck in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant rat Lck is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction