Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human LDLR Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB2148S100UG
This item is not returnable.
View return policy
Description
LDLR Monoclonal specifically detects LDLR in Human samples. It is validated for Flow Cytometry.Specifications
LDLR | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P01130 | |
LDLR | |
Chinese hamster ovary cell line CHO-derived recombinant human LDL R Ala22-Arg788 Accession # P01130 | |
RUO | |
3949 | |
Human | |
IgG1 |
Flow Cytometry | |
472413 | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
FH, FHC, LDL R, LDL receptor, LDLCQ2, low density lipoprotein receptor, low-density lipoprotein receptor, low-density lipoprotein receptor class A domain-containing protein 3 | |
Mouse | |
100 μg | |
Primary | |
Detects human LDL R in ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) LDL R, recombinant human LRP-5, or rmLRP-6 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction