Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human LDLR Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 19 publications
Supplier: R&D Systems AF2148SP
This item is not returnable.
View return policy
Description
LDLR Polyclonal specifically detects LDLR in Human samples. It is validated for Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction, Immunocytochemistry.Specifications
LDLR | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FH, FHC, LDL R, LDL receptor, LDLCQ2, low density lipoprotein receptor, low-density lipoprotein receptor, low-density lipoprotein receptor class A domain-containing protein 3 | |
Goat | |
Affinity Purified | |
RUO | |
3949 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Blockade of Receptor-ligand Interaction | |
P01130 | |
LDLR | |
Chinese hamster ovary cell line CHO-derived recombinant human LDL R Asp193-Arg788 Accession # P01130 | |
25 μg | |
Primary | |
Detects human LDL R in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant mouse LDL R is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction