Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human LECT2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF722
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF722 100 μg
AF722SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF722 Supplier R&D Systems Supplier No. AF722
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

LECT2 Polyclonal specifically detects LECT2 in Human samples. It is validated for Western Blot.

Specifications

Antigen LECT2
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Gene Accession No. O14960
Gene Alias chm2, chm-II, Chondromodulin II, chondromodulin-II, hLECT2, LECT-2, leukocyte cell-derived chemotaxin 2, leukocyte cell-derived chemotaxin-2, MGC126628, MIM1
Gene Symbols LECT2
Host Species Goat
Immunogen E. coli-derived recombinant human LECT2 Gly19-Leu151 Accession # O14960
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3950
Test Specificity Detects human LECT2 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.