Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human LEDGF Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF3468SP
This item is not returnable.
View return policy
Description
LEDGF Polyclonal specifically detects LEDGF in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry, Knockout Validated.Specifications
LEDGF | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CLL-associated antigen KW-7, Dense fine speckles 70 kDa protein, DFS 70, DFS70, LEDGFDFS 70, Lens epithelium-derived growth factor, MGC74712, p52, p75, PAIP, PC4 and SFRS1 interacting protein 1, PC4 and SFRS1 interacting protein 2, PC4 and SFRS1-interacting protein, PSIP1, PSIP2, transcriptional coactivator p52/p75, Transcriptional coactivator p75/p52 | |
Goat | |
Affinity Purified | |
RUO | |
11168 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunocytochemistry, Immunoassay | |
LYOPH | |
Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Immunocytochemistry 5-15 ug/mL, Knockout Validated | |
O75475 | |
PSIP1 | |
E. coli-derived recombinant human LEDGF Met1-Asn530 Accession # O75475 | |
25 μg | |
Primary | |
Detects human LEDGF in direct ELISAs and Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction