Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human LEDGF Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3468
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3468 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3468 Supplier R&D Systems Supplier No. AF3468
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

LEDGF Polyclonal specifically detects LEDGF in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry, Knockout Validated.

Specifications

Antigen LEDGF
Applications Western Blot, Immunocytochemistry, Immunoassay
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Immunocytochemistry 5-15 ug/mL, Knockout Validated
Gene Accession No. O75475
Gene Alias CLL-associated antigen KW-7, Dense fine speckles 70 kDa protein, DFS 70, DFS70, LEDGFDFS 70, Lens epithelium-derived growth factor, MGC74712, p52, p75, PAIP, PC4 and SFRS1 interacting protein 1, PC4 and SFRS1 interacting protein 2, PC4 and SFRS1-interacting protein, PSIP1, PSIP2, transcriptional coactivator p52/p75, Transcriptional coactivator p75/p52
Gene Symbols PSIP1
Host Species Goat
Immunogen E. coli-derived recombinant human LEDGF Met1-Asn530 Accession # O75475
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11168
Test Specificity Detects human LEDGF in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.