Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human LEF1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF7647
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF7647 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF7647 Supplier R&D Systems Supplier No. AF7647
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 2 publications

LEF1 Polyclonal specifically detects LEF1 in Human samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen LEF1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.2 ug/mL, Immunocytochemistry 1-15 ug/mL
Gene Accession No. Q9UJU2
Gene Alias DKFZp586H0919, FLJ46390, LEF-1, lymphoid enhancer-binding factor 1, T cell-specific transcription factor 1-alpha, TCF10, TCF1-alpha, TCF1ALPHA, TCF7L3
Gene Symbols LEF1
Host Species Goat
Immunogen E. coli-derived recombinant human LEF1 Met120-Met280 (predicted) Accession # Q9UJU2
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51176
Test Specificity Detects human LEF1 in direct ELISAs and Western blots. In direct ELISAs, approximately 35% cross-reactivity with recombinant human (rh) TCF-1, rhTCF-2, and rhTCF-4 is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.