Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Human Lefty-A ELISA Kit
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
Description
Human Lefty-A quantitates human Lefty-A in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Lefty-A.
Left-right determination factor 1 and 2 (Lefty1/2) belong to the TGF-beta superfamily of proteins and are responsible for determination of left-right positioning of visceral organs during embryogenesis. Lefty1 acts as a regulator of Lefty2 and NODAL as the lack of the former resulted in bilateral expression of nodal, lefty2 and Pitx2 (a homeobox gene normally expressed on the left side and blocks the signal for leftness). Lefty2 may play a role in endometrial bleeding.
Specifications
Specifications
| Accession Number | 6030463A22Rik; AV214969; EBAF; Ebaf2; endometrial bleeding associated factor; endometrial bleeding-associated factor; LEFTA; Leftb; Left-right determination factor 2; left-right determination factor A; Left-right determination factor B; left-right determination, factor A; Lefty2; LEFTYA; MGC46222; Protein lefty-2; protein lefty-A; Protein lefty-B; PSEC0024; TGFB4; TGF-beta-4; Transforming growth factor beta-4 |
| Assay Range | 0.82 to 200 ng/mL |
| Assay Sensitivity | 0.82 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 7044 |
| Gene Symbol | LEFTY2 |
| Interassay CV | <12% |
| Show More |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction