Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Lefty-A Long Isoform Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB746
This item is not returnable.
View return policy
Description
Lefty-A Monoclonal specifically detects Lefty-A in Human samples. It is validated for Western Blot.Specifications
Lefty-A | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EBAFTGF-beta-4, endometrial bleeding associated factor, Endometrial bleeding-associated factor, Lefta, LEFTAProtein lefty-A, left-right determination factor 2, Left-right determination factor A, LEFTY2, LeftyA, LEFTYATransforming growth factor beta-4, MGC46222, Protein lefty-2, TGFB4endometrial bleeding associated factor (left-right determination, factor A;transforming growth factor beta superfamily) | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
7044 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Western Blot | |
182515 | |
Western Blot 1 ug/mL | |
O00292.2 | |
LEFTY2 | |
Mouse myeloma cell line NS0-derived recombinant human Lefty-A Phe78-Pro366 Accession # O00292.2 | |
500 μg | |
Primary | |
Detects the long isoform of recombinant human (rh) Lefty-A (aa 78-366) in direct ELISAs and Western blots. Does not cross-react with the short form of recombinant human Lefty-A (aa 136-366) or with rhART, recombinant mouse (rm) Artemin, recombinant drosophila DPP, rhGDNF, rhLAP, recombinant rat MIS, rhNeurturin, rhTGF-alpha , rhTGF-beta 1, rhTGF-beta 1.2, or rmLefty-1. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction