Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Legumain/Asparaginyl Endopeptidase Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2199
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2199 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB2199 Supplier R&D Systems Supplier No. MAB2199
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Legumain/Asparaginyl Endopeptidase Monoclonal specifically detects Legumain/Asparaginyl Endopeptidase in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Legumain/Asparaginyl Endopeptidase
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 312114
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q99538
Gene Alias AEP, Asparaginyl endopeptidase, cysteine protease 1, legumain, LGMN, LGMN1, Protease, cysteine 1, protease, cysteine, 1 (legumain), PRSC1EC 3.4.22.34
Gene Symbols LGMN
Host Species Mouse
Immunogen NS0-derived recombinant human Legumain/Asparaginyl Endopeptidase Ile18-Tyr433 Accession # Q99538
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5641
Test Specificity Detects human Legumain/Asparaginyl Endopeptidase (aa18-433) in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse Legumain is observed. Recognizes the pro form of human Legumain and does not recognize the mature form (aa 18-323).
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.