Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Legumain/Asparaginyl Endopeptidase Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB21992
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB21992 500 μg
MAB21992SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB21992 Supplier R&D Systems Supplier No. MAB21992
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Legumain/Asparaginyl Endopeptidase Monoclonal specifically detects Legumain/Asparaginyl Endopeptidase in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen Legumain/Asparaginyl Endopeptidase
Applications Western Blot, ELISA
Classification Monoclonal
Clone 312109
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q99538
Gene Alias AEP, Asparaginyl endopeptidase, cysteine protease 1, legumain, LGMN, LGMN1, Protease, cysteine 1, protease, cysteine, 1 (legumain), PRSC1EC 3.4.22.34
Gene Symbols LGMN
Host Species Mouse
Immunogen NS0-derived recombinant human Legumain Val18-Tyr433 Accession # Q99538
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5641
Test Specificity Detects both the pro and mature forms of human Legumain/Asparaginyl Endopeptidase in ELISAs and Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.