Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Leptin R Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

Supplier:  R&D Systems FAB867G100UG

Catalog No. FAB867G100


Only null left
Add to Cart
This item is not returnable. View return policy

Description

Description

Leptin R Monoclonal specifically detects Leptin R in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Leptin R
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
P48357
LEPR
Mouse myeloma cell line NS0-derived recombinant human Leptin R Phe22-Asp839 (predicted) Accession # P48357
RUO
3953
Human
IgG2b
Flow Cytometry
52263
Alexa Fluor 488
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
B219, CD295, CD295 antigen, DB, DKFZp686B1731, huB219, LEPR, LEP-R, leptin receptor, LeptinR, OB R, OB receptor, OB-R, OBRCD295
Mouse
100 μg
Primary
Detects human Leptin R in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant mouse Leptin R is observed.
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only (RUO)