Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Leptin R Antibody, Novus Biologicals™

Mouse Monoclonal Antibody
$177.10 - $433.50
Specifications
Antigen | Leptin R |
---|---|
Clone | 52276 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB86713100
![]() |
R&D Systems
MAB86713100 |
100 μg |
Each for $433.50
|
|
|||||
MAB86713SP
![]() |
R&D Systems
MAB86713SP |
25 μg |
Each for $177.10
|
|
|||||
Description
Leptin R Monoclonal antibody specifically detects Leptin R in nan samples. It is validated for ELISASpecifications
Leptin R | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
P48357 | |
3953 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
52276 | |
ELISA | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
B219, CD295, CD295 antigen, DB, DKFZp686B1731, huB219, LEPR, LEP-R, leptin receptor, LeptinR, OB R, OB receptor, OB-R, OBRCD295 | |
Mouse myeloma cell line NS0-derived recombinant human Leptin R, Accession # P48357 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title