Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Leptin R Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB867100
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB867100 100 μg
MAB867SP 25 μg
MAB867500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB867100 Supplier R&D Systems Supplier No. MAB867100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 4 publications

Leptin R Monoclonal specifically detects Leptin R in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready.

Specifications

Antigen Leptin R
Applications Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF
Classification Monoclonal
Clone 52263
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P48357
Gene Alias B219, CD295, CD295 antigen, DB, DKFZp686B1731, huB219, LEPR, LEP-R, leptin receptor, LeptinR, OB R, OB receptor, OB-R, OBRCD295
Gene Symbols LEPR
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Leptin R Phe22-Asp839 (predicted) Accession # P48357
Purification Method Protein A or G purified from ascites
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3953
Test Specificity Detects human Leptin R in direct ELISAs and Western blots. In Western blots, cross reactivity with recombinant mouse Leptin R is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Ascites
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.