Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Leptin R MAb (Clone 52209), R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $491.00
Specifications
| Antigen | Leptin R |
|---|---|
| Clone | 52209 |
| Dilution | Neutralization 0.2-1 ug/mL |
| Applications | Neutralization |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB86712SP
![]() |
R&D Systems
MAB86712SP |
25 μg |
Each for $200.50
|
|
|||||
MAB86712100
![]() |
R&D Systems
MAB86712100 |
100 μg |
Each for $491.00
|
|
|||||
Description
Leptin R Monoclonal specifically detects Leptin R in Human samples. It is validated for Neutralization.Specifications
| Leptin R | |
| Neutralization 0.2-1 ug/mL | |
| Monoclonal | |
| Supernatant | |
| Human | |
| P48357 | |
| 3953.0 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| 52209 | |
| Neutralization | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| B219, CD295, CD295 antigen, DB, DKFZp686B1731, huB219, LEPR, LEP-R, leptin receptor, LeptinR, OB R, OB receptor, OB-R, OBRCD295 | |
| Mouse myeloma cell line NS0-derived recombinant human Leptin R, Phe22-Asp839, Accession # P48357 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Leptin R in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title