Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Leptin R PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB867P
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB867P 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB867P Supplier R&D Systems Supplier No. FAB867P
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Leptin R Monoclonal specifically detects Leptin R in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Leptin R
Applications Flow Cytometry
Classification Monoclonal
Clone 52263
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. P48357
Gene Alias B219, CD295, CD295 antigen, DB, DKFZp686B1731, huB219, LEPR, LEP-R, leptin receptor, LeptinR, OB R, OB receptor, OB-R, OBRCD295
Gene Symbols LEPR
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Leptin R Phe22-Asp839 (predicted) Accession # P48357
Purification Method Protein A or G purified from ascites
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3953
Test Specificity Detects human Leptin R in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant mouse Leptin R is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.