Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Leucyl-cystinyl Aminopeptidase/LNPEP Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF6386
This item is not returnable.
View return policy
Description
Leucyl-cystinyl Aminopeptidase/LNPEP Polyclonal specifically detects Leucyl-cystinyl Aminopeptidase/LNPEP in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Leucyl-cystinyl Aminopeptidase/LNPEP | |
Polyclonal | |
Unconjugated | |
Q9UIQ6 | |
LNPEP | |
Chinese hamster ovary cell line CHO-derived recombinant human Leucyl-cystinyl Aminopeptidase/LNPEP Arg133-Gly552 Accession # Q9UIQ6 | |
100 μg | |
Primary | |
Detects human Leucyl-cystinyl Aminopeptidase/LNPEP in direct ELISAs and Western blots. In direct ELISAs, less than 2% cross-reactivity with recombinant human (rh) ENPEP, rhLRAP, and rhARTS1 is observed. | |
Human | |
IgG |
Western Blot, Immunoprecipitation | |
LYOPH | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
angiotensin IV receptor, AT (4) receptor, CAP, Cystinyl aminopeptidase, EC 3.4.11, EC 3.4.11.3, Insulin-regulated membrane aminopeptidase, Insulin-responsive aminopeptidase, IRAP, leucyl/cystinyl aminopeptidase, leucyl-cystinyl aminopeptidase, Leucylcystinyl Aminopeptidase, LNPEP, Otase, Oxytocinase, Placental leucine aminopeptidase, PLAP, P-LAP, P-LAPinsulin-regulated aminopeptidase, vasopressinase | |
Sheep | |
Affinity Purified | |
RUO | |
4012 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction