Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Leukotriene B4 R1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB099SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB099SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB099SP Supplier R&D Systems Supplier No. MAB099SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

Leukotriene B4 R1 Monoclonal specifically detects Leukotriene B4 R1 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen Leukotriene B4 R1
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 203/14F11
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias BLT1, chemokine receptor-like 1, CMKRL1, leukotriene B4 receptor, Leukotriene B4R1, LTB4R, LTB4R1, LTBR1, P2Y purinoceptor 7, purinergic receptor P2Y, G-protein coupled, 7
Gene Symbols LTB4R
Host Species Mouse
Immunogen HeLa cervical epithelial carcinoma cell line transfected with human Leukotriene B4 R1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1241
Test Specificity Recognizes human Leukotriene B4 R1 (Pettersson, A. et al. (2000) Biochem. Biophys. Res. Commun. 279:520).
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.