Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Leukotriene B4 R1 PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB099P
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB099P 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB099P Supplier R&D Systems Supplier No. FAB099P
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 3 publications

Leukotriene B4 R1 Monoclonal specifically detects Leukotriene B4 R1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Leukotriene B4 R1
Applications Flow Cytometry
Classification Monoclonal
Clone 203/14F11
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Alias BLT1, chemokine receptor-like 1, CMKRL1, leukotriene B4 receptor, Leukotriene B4R1, LTB4R, LTB4R1, LTBR1, P2Y purinoceptor 7, purinergic receptor P2Y, G-protein coupled, 7
Gene Symbols LTB4R
Host Species Mouse
Immunogen HeLa cervical epithelial carcinoma cell line transfected with human Leukotriene B4 R1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1241
Test Specificity Recognizes human Leukotriene B4 R1 (5).
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.