Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Lgr4/GPR48 PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB7750P025
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB7750P025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB7750P025 Supplier R&D Systems Supplier No. FAB7750P025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Lgr4/GPR48 Monoclonal specifically detects Lgr4/GPR48 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Lgr4/GPR48
Applications Flow Cytometry
Classification Monoclonal
Clone 852229
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q9BXB1
Gene Alias GPR48, GPR48G protein-coupled receptor 48, G-protein coupled receptor 48, leucine-rich repeat containing G protein-coupled receptor 4
Gene Symbols LGR4
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human Lgr4/GPR48 Met1-Asp951 Accession # Q9BXB1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55366
Test Specificity Detects human Lgr4/GPR48 in Lgr4-transfected cell-based ELISA. Stains human Lgr4/GPR48 transfected but not irrelevant transfected cells in flow cytometry.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.