Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Lgr6 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB84581V100UG
This item is not returnable.
View return policy
Description
Lgr6 Monoclonal specifically detects Lgr6 in Human samples. It is validated for Flow Cytometry.Specifications
Lgr6 | |
Monoclonal | |
Alexa Fluor 405 | |
Supplied 0.2Mg/mL in a saline solution containing BSA and Sodium Azide. | |
FLJ14471, gonadotropin receptor, GPCR, leucine-rich repeat containing G protein-coupled receptor 6, leucine-rich repeat-containing G protein-coupled receptor 6, leucine-rich repeat-containing G-protein coupled receptor 6, VTS20631 | |
NS0 mouse myeloma cell line transfected with human Lgr6 Met1-Val967 Accession # Q9HBX8 | |
100 μg | |
59352 | |
Human | |
Supernatant |
Flow Cytometry | |
918726 | |
Flow Cytometry 0.25-1μg/106 cells | |
Q9HBX8 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human Lgr6 in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction