Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human LIF, Animal-Free Recombinant Protein, PeproTech®

Catalog No. 50398584
Encompass_Preferred
Click to view available options
Quantity:
250 μg

Recombinant Protein

Recombinant Human LIF is a 19.7 kDa protein containing 180 amino acid residues, including three disulfide bonds. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Leukemia inhibitory factor (LIF) is a 20 kDa protein that belongs to the IL-6 receptor family. It binds to a heterodimeric membrane receptor made up of a LIF-specific subunit, gp190 or LIFR, and the subunit gp130, which is shared with the other members of the IL-6 family. LIF expression has been observed in various tissues including thymus, lung, and neuronal tissue. LIF can be up-regulated by pro-inflammatory cytokines such as TNFα and IL-17, and elevated levels of LIF have been found in cases of rheumatoid arthritis, neural injury, systemic inflammation, and tuberculosis. LIF displays diverse biological effects, but is best known for its ability to inhibit the differentiation of embryonic stem cells in mice and contribute to stem cell self-renewal.Human and mouse LIF share 79% sequence homology and exhibit cross-species activity. However, LIF inhibition of stem cell differentiation appears to be mouse-specific.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P15018
For Use With (Application) Functional Assay
Format Lyophilized
Formulation protein with no preservative
Gene ID (Entrez) 3976
Molecular Weight (g/mol) 19.6 kDa
Name Human LIF, Animal-Free
Purification Method HPLC, SDS-PAGE
Quantity 250 μg
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/ μg
Gene Alias CDF; cholinergic differentiation factor; cholinergic neuronal differentiation factor; d factor; DIA; differentiation inhibitory activity; differentiation-inducing factor; differentiation-stimulating factor; Emfilermin; hepatocyte-stimulating factor III; HILDA; human interleukin in DA cells; leukemia inhibitory factor; leukemia inhibitory factor (cholinergic differentiation factor); LIF; Melanoma-derived LPL inhibitor; MLPLI; myeloid leukaemia inhibitory factor
Common Name LIF
Gene Symbol Lif
Biological Activity Determined by its ability to stimulate the proliferation of human TF-1 cells. The expected ED50 is ≤ 0.1 ng/ml, corresponding to a specific activity of ≥ 1 x 10^7 units/mg
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence SPLPITPVNA TCAIRHPCHN NLMNQIRSQL AQLNGSANAL FILYYTAQGE PFPNNLDKLC GPNVTDFPPF HANGTEKAKL VELYRIVVYL GTSLGNITRD QKILNPSALS LHSKLNATAD ILRGLLSNVL CRLCSKYHVG HVDVTYGPDT SGKDVFQKKK LGCQLLGKYK QIIAVLAQAF
Content And Storage -20°C
Activity Determined by its ability to stimulate the proliferation of human TF-1 cells. The expected ED50 is ≤ 0.1 ng/ml, corresponding to a specific activity of ≥ 1 x 10^7 units/mg
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.