Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human LIF Antibody, R&D Systems™

Mouse Monoclonal Antibody
$188.70 - $474.50
Specifications
Antigen | LIF |
---|---|
Clone | 945842 |
Applications | ELISA |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB2502100
![]() |
R&D Systems
MAB2502100 |
100 μg |
Each for $474.50
|
|
|||||
MAB2502SP
![]() |
R&D Systems
MAB2502SP |
25 μg |
Each for $188.70
|
|
|||||
Description
LIF Monoclonal antibody specifically detects LIF in Human samples. It is validated for ELISASpecifications
LIF | |
ELISA | |
Unconjugated | |
Mouse | |
Human | |
P15018 | |
3976 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. |
945842 | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
CDF, D factor, DIA, differentiation inhibitory activity, differentiation stimulating factor, Differentiation-stimulating factor, Emfilermin, HILDAcholinergic differentiation factor, leukemia inhibitory factor, leukemia inhibitory factor (cholinergic differentiation factor), Melanoma-derived LPL inhibitor, MLPLI | |
Human embryonic kidney cell, HEK293-derived human LIF, Met1-Phe202, Accession # P15018 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title