Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human LIF Quantikine ELISA Kit

Catalog No. dlf00b
Click to view available options
Quantity:
1 Kit
This item is not returnable. View return policy
This item is not returnable. View return policy

From the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

  • A 4.5 hours solid phase ELISA designed to measure human LIF in cell culture supernates, serum, and plasma.
  • It contains HEK293-expressed recombinant human LIF and antibodies raised against the recombinant protein.
  • Results obtained for naturally occurring human LIF showed linear curves that were parallel to the standard curves obtained using the recombinant Quantikine™ kit standards.
  • These results indicate that this kit can be used to determine relative mass values for natural human LIF.
  • Intra-Assay Precision (Precision within an assay)Three samples of known concentration were tested twenty times on one plate to assess intra-assay precision.
  • Inter-Assay Precision (Precision between assays) Three samples of known concentration were tested in twenty separate assays to assess inter-assay precision. Assays were performed by at least three technicians using two lots of components.

Specifications

Product Type ELISA
Specificity Natural and Recombinant Human LIF
Gene ID (Entrez) 3976
Host Species Human
Quantity 1 Kit
Storage Requirements Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Synonym CDF, D factor, DIA, differentiation inhibitory activity, differentiation stimulating factor, Differentiation-stimulating factor, Emfilermin, HILDA, HILDAcholinergic differentiation factor, leukemia inhibitory factor, leukemia inhibitory factor (cholinergic differentiation factor), Melanoma-derived LPL inhibitor, MLPLI
Target LIF
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.