Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human LIGHT/TNFSF14 Alexa Fluor 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB664T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB664T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB664T100 Supplier R&D Systems Supplier No. FAB664T100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

LIGHT/TNFSF14 Monoclonal specifically detects LIGHT/TNFSF14 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen LIGHT/TNFSF14
Applications Flow Cytometry
Classification Monoclonal
Clone 115520
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 594
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. O43557
Gene Alias CD258, CD258 antigen, delta transmembrane LIGHT, Herpes virus entry mediator ligand, Herpesvirus entry mediator ligand, HVEM-L, HVEMLherpesvirus entry mediator-ligand, ligand for herpesvirus entry mediator, LIGHTherpesvirus entry mediator A, LTg, TNFSF14, TR2, tumor necrosis factor (ligand) superfamily, member 14, tumor necrosis factor ligand superfamily member 14, tumor necrosis factor receptor-like 2, tumor necrosis factor superfamily member LIGHT
Gene Symbols TNFSF14
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human LIGHT/TNFSF14 Asp74-Val240 Accession # O43557
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8740
Test Specificity Detects human LIGHT/TNFSF14 in ELISAs. In sandwich immunoassays, no significant cross-reactivity or interference with recombinant human (rh) Fas Ligand, rhAPRIL, recombinant mouse TRANCE, rhTNF-alpha or rhTRAIL is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.