Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human LIGHT/TNFSF14 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6643100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6643100 100 μg
MAB6643SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB6643100 Supplier R&D Systems Supplier No. MAB6643100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

LIGHT/TNFSF14 Monoclonal specifically detects LIGHT/TNFSF14 in Human samples. It is validated for Flow Cytometry, Neutralization, CyTOF-ready.

Specifications

Antigen LIGHT/TNFSF14
Applications Flow Cytometry, Neutralization, CyTOF
Classification Monoclonal
Clone 2323C
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 μg/106 cells, Neutralization 6-48 ng/mL, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. O43557
Gene Alias CD258, CD258 antigen, delta transmembrane LIGHT, Herpes virus entry mediator ligand, Herpesvirus entry mediator ligand, HVEM-L, HVEMLherpesvirus entry mediator-ligand, ligand for herpesvirus entry mediator, LIGHTherpesvirus entry mediator A, LTg, TNFSF14, TR2, tumor necrosis factor (ligand) superfamily, member 14, tumor necrosis factor ligand superfamily member 14, tumor necrosis factor receptor-like 2, tumor necrosis factor superfamily member LIGHT
Host Species Rabbit
Immunogen Mouse myeloma cell line NS0-derived recombinant human LIGHT/TNFSF14, Asp74-Val240, Accession # O43557
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 8740
Test Specificity Detects human LIGHT/TNFSF14 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.