Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human LILRA1/CD85i/LIR-6 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB30851N100UG
This item is not returnable.
View return policy
Description
LIR-6/LILRA1 Monoclonal specifically detects LIR-6/LILRA1 in Human samples. It is validated for Flow Cytometry.Specifications
LIR-6/LILRA1 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
O75019 | |
LILRA1 | |
Mouse myeloma cell line NS0-derived recombinant human LILRA1/CD85i/LIR-6 Pro17-Asn461 Accession # O75019 | |
RUO | |
11024 | |
Human | |
IgG2b |
Flow Cytometry | |
586326 | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CD85 antigen-like family member I, CD85i, CD85i antigen, leukocyte immunoglobulin-like receptor subfamily A member 1, leukocyte immunoglobulin-like receptor subfamily A member 1 soluble isoform, leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 1, LIR6, LIR-6, LIR-6Leukocyte immunoglobulin-like receptor 6, LIR6MGC126563 | |
Mouse | |
100 μg | |
Primary | |
Detects human LILRA1/CD85i/LIR-6 in direct ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant human (rh) ILT2 is observed and no cross-reactivity rhILT3, 4, 5, 6, rhLIR-7 or -8 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction