Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human LILRA1/CD85i/LIR-6 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB30852SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB30852SP 25 μg
MAB30852100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB30852SP Supplier R&D Systems Supplier No. MAB30852SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

LIR-6/LILRA1 Monoclonal specifically detects LIR-6/LILRA1 in Human samples. It is validated for Immunocytochemistry.

Specifications

Antigen LIR-6/LILRA1
Applications Immunocytochemistry
Classification Monoclonal
Clone 586439
Conjugate Unconjugated
Dilution Immunocytochemistry 8-25 μ/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. O75019
Gene Alias CD85 antigen-like family member I, CD85i, CD85i antigen, leukocyte immunoglobulin-like receptor subfamily A member 1, leukocyte immunoglobulin-like receptor subfamily A member 1 soluble isoform, leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 1, LIR6, LIR-6, LIR-6Leukocyte immunoglobulin-like receptor 6, LIR6MGC126563
Host Species Mouse
Immunogen Mouse myeloma cell line, NS0-derived human LILRA1/CD85i/LIR-6, Pro17-Asn461, Accession No. O75019
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 11024
Test Specificity Detects human LILRA1/CD85i/LIR-6 in direct ELISAs. In direct ELISAs, no cross-reactivity with human LILRA2, LILRA3, LILRA4, LILRA5, LILRA6, LILRB2 and LILRB3 was observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.