Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human LILRA1/CD85i/LIR-6 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB3085
This item is not returnable.
View return policy
Description
LIR-6/LILRA1 Monoclonal specifically detects LIR-6/LILRA1 in Human samples. It is validated for Western Blot.Specifications
| LIR-6/LILRA1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| O75019 | |
| LILRA1 | |
| Mouse myeloma cell line NS0-derived recombinant human LILRA1/CD85i/LIR-6 Pro17-Asn461 Accession # O75019 | |
| 100 μg | |
| Primary | |
| Detects human LILRA1/CD85i/LIR-6 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) ILT2, rhILT4, rhILT5, rhLIR-4, rhLIR-5, or rhLIR-8 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 356316 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD85 antigen-like family member I, CD85i, CD85i antigen, leukocyte immunoglobulin-like receptor subfamily A member 1, leukocyte immunoglobulin-like receptor subfamily A member 1 soluble isoform, leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 1, LIR6, LIR-6, LIR-6Leukocyte immunoglobulin-like receptor 6, LIR6MGC126563 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 11024 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction