Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human LILRA1/LILRB1 APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB30851A
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB30851A 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB30851A Supplier R&D Systems Supplier No. FAB30851A
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

LILRA1/LILRB1 Monoclonal specifically detects LILRA1/LILRB1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen LILRA1/LILRB1
Applications Flow Cytometry
Classification Monoclonal
Clone 586326
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. O75019
Gene Alias CD85I, CD85j, ILT2, LIR1, LIR6, LIR-6, MIR7
Gene Symbols LILRA1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human LILRA1 Pro17-Asn461 Accession # O75019
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11024
Test Specificity Detects human LILRA1 in direct ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant human (rh) LILRB1 is observed and no cross-reactivity rhILT3, 4, 5, 6, rhLIR-7 or -8 is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.