Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human LILRA4/CD85g/ILT7 Alexa Fluor 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB6287N100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB6287N100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB6287N100 Supplier R&D Systems Supplier No. FAB6287N100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ILT7/CD85g/LILRA4 Monoclonal specifically detects ILT7/CD85g/LILRA4 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen ILT7/CD85g/LILRA4
Applications Flow Cytometry
Classification Monoclonal
Clone 656688
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 700
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P59901
Gene Alias CD85 antigen-like family member G, CD85g, CD85g antigen, ILT7, ILT-7, ILT7MGC129598, Immunoglobulin-like transcript 7, leukocyte immunoglobulin-like receptor subfamily A member 4, leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 4, leukocyte immunoglobulin-like receptor, subfamily A (without TM domain), member4, MGC129597
Gene Symbols LILRA4
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human LILRA4/CD85g/ILT7 Glu24-Asn446 Accession # P59901
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23547
Test Specificity Detects human LILRA4/CD85g/ILT7 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) LILRA5, rhILT2, 3, 4, 5, or 6 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.