Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human LILRA5/CD85f Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB6754N100UG
This item is not returnable.
View return policy
Description
ILT11/LILRA5 Monoclonal specifically detects ILT11/LILRA5 in Human samples. It is validated for Flow Cytometry.Specifications
ILT11/LILRA5 | |
Monoclonal | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CD85, CD85 antigen-like family member F, CD85F, CD85f antigen, ILT11, ILT-11, ILT11CD85f, Immunoglobulin-like transcript 11, leukocyte Ig-like receptor 9, Leukocyte immunoglobulin-like receptor 9, leukocyte immunoglobulin-like receptor subfamily A member 5, leukocyte immunoglobulin-like receptor subfamily A member 5 soluble, leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 5, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LILRB7, LIR9, LIR-9, LIR9immunoglobulin-like transcript 11 protein, member 7 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human ILT11/LILRA5 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse ILT11/LILRA5, recombinant rat (rr) ILT11/LILRA5, rrLILRC2, recombinant human (rh) ILT4/CD85d, rhILT5/CD85a, or rhILT3/CD85k is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG2a |
Flow Cytometry | |
711828 | |
Flow Cytometry 0.25-1 μg/106 cells | |
A6NI73 | |
LILRA5 | |
Chinese hamster ovary cell line CHO-derived recombinant human ILT11/LILRA5, Gly42-Arg268, Accession # A6NI73 | |
100 μg | |
353514 | |
Human | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction