Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human LILRA6/CD85b APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB8656A025
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB8656A025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB8656A025 Supplier R&D Systems Supplier No. FAB8656A025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ILT8/CD85b/LILRA6 Monoclonal specifically detects ILT8/CD85b/LILRA6 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen ILT8/CD85b/LILRA6
Applications Flow Cytometry
Classification Monoclonal
Clone 921330
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q6PI73
Gene Symbols LILRA6
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line-derived recombinant human LILRA6/CD85b Met1-Asn447 Accession # Q6PI73
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79168
Test Specificity Detects human LILRA6/CD85b in direct ELISA. Detects HEK293 cells transfected with LILRA6/CD85b but not irrelevant transfectants.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.