Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human LILRA6 (CD85b, ILT8) Fc Chimera Recombinant Protein

Catalog No. PIRP88121
Encompass
Click to view available options
Quantity:
1 mg
100 μg

Recombinant Protein

It is recommended that the protein be aliquoted for optimal storage. It is recommended that sterile water (1785.71428571429 μL) be added to the vial to prepare a stock solution of 0.56 mg/mL. Concentration is measured by UV.

LILRA6 may act as receptor for class I MHC antigens.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q6PI73
For Use With (Application) Control
Format Lyophilized
Formulation PBS with 0.01% mannitol, 0.01% Tween 80, 5-8% trehalose and no preservative; pH 7.4
Gene ID (Entrez) 79168
Molecular Weight (g/mol) 89 kDa
Name Human LILRA6 (CD85b, ILT8) Fc Chimera
Purification Method SDS-PAGE
Quantity 100 μg
Regulatory Status RUO
Endotoxin Concentration <1.0 EU/μg
Gene Alias CD85b; ILT5; ILT8; ILT-8; immunoglobulin-like transcript 5; immunoglobulin-like transcript 8; leucocyte Ig-like receptor A6; leukocyte Ig-like receptor; leukocyte immunoglobulin like receptor A6; leukocyte immunoglobulin-like receptor subfamily A member 6; leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 6; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 6; LILRA6; LILRB3; LILRB6
Common Name LILRA6
Gene Symbol LILRA6
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag Fc-tag
Sequence Human LILRA6, amino acids Met1-Asn447 (Accession # Q6PI73-1) with a C-terminal human IgG1 Fc region
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade >90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.