Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human LILRB1/CD85j/ILT2 Alexa Fluor 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FA20171R100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FA20171R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FA20171R100 Supplier R&D Systems Supplier No. FAB20171R100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ILT2/CD85j/LILRB1 Monoclonal specifically detects ILT2/CD85j/LILRB1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen ILT2/CD85j/LILRB1
Applications Flow Cytometry
Classification Monoclonal
Clone 292305
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 647
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q8NHL6
Gene Alias CD85 antigen-like family member J, CD85J, CD85j antigen, Ig-like transcript 2, ILT2, ILT-2, ILT2FLJ37515, Immunoglobulin-like transcript 2, leukocyte Ig-like receptor-1, Leukocyte immunoglobulin-like receptor 1, leukocyte immunoglobulin-like receptor subfamily B member 1, leukocyte immunoglobulin-like receptor subfamily B member 1 soluble isoform, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR1, LIR1LIR-1MIR-7CD85MIR7CD85 antigen, member 1, MIR7, Monocyte/macrophage immunoglobulin-like receptor 7
Gene Symbols LILRA5
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human LILRB1/CD85j/ILT2 Gly24-His458 Accession # Q8NHL6
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10859
Test Specificity Detects human LILRB1/CD85j/ILT2 in Western blots. In Western blots, no cross-reactivity with recombinant human (rh) ILT4, rhILT5, rhLIR6 or rhLIR8 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.