Learn More
Human LILRB1/CD85j/ILT2 PE-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | ILT2/CD85j/LILRB1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 292305 |
| Conjugate | R-PE |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Formulation | Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide |
| Gene Accession No. | Q8NHL6 |
| Gene Alias | CD85 antigen-like family member J, CD85J, CD85j antigen, Ig-like transcript 2, ILT2, ILT-2, ILT2FLJ37515, Immunoglobulin-like transcript 2, leukocyte Ig-like receptor-1, Leukocyte immunoglobulin-like receptor 1, leukocyte immunoglobulin-like receptor subfamily B member 1, leukocyte immunoglobulin-like receptor subfamily B member 1 soluble isoform, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR1, LIR1LIR-1MIR-7CD85MIR7CD85 antigen, member 1, MIR7, Monocyte/macrophage immunoglobulin-like receptor 7 |
| Gene Symbols | LILRB1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.