Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human LILRB2/CD85d/ILT4 Alexa Fluor 350-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB2078U100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB2078U100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB2078U100 Supplier R&D Systems Supplier No. FAB2078U100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ILT4/CD85d/LILRB2 Monoclonal specifically detects ILT4/CD85d/LILRB2 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen ILT4/CD85d/LILRB2
Applications Flow Cytometry
Classification Monoclonal
Clone 287219
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 350
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. ACT64556
Gene Alias CD85 antigen-like family member D, CD85d, CD85d antigen, Ig-like transcript 4, ILT4, ILT-4, ILT4CD85d, Immunoglobulin-like transcript 4, Leukocyte immunoglobulin-like receptor 2, leukocyte immunoglobulin-like receptor subfamily B member 2, leukocyte immunoglobulin-like receptor subfamily B member 2 soluble isoform, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR2, LIR2CD85D, LIR-2subfamily A (with TM domain), member 6, member 2, MIR10, MIR10LILRA6, Monocyte/macrophage immunoglobulin-like receptor 10
Gene Symbols LILRB2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human LILRB2/CD85d/ILT4 Gly24-His458 Accession # ACT64556
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10288
Test Specificity Detects human LILRB2/CD85d/ILT4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) ILT1, rhILT2, rhILT3, rhILT6, rhILT7, rhILT11, rhLIR6 or rhLIR8 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.