Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human LILRB2/CD85d/ILT4 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB2078R100UG
This item is not returnable.
View return policy
Description
ILT4/CD85d/LILRB2 Monoclonal specifically detects ILT4/CD85d/LILRB2 in Human samples. It is validated for Flow Cytometry.Specifications
ILT4/CD85d/LILRB2 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
ACT64556 | |
LILRB2 | |
Mouse myeloma cell line NS0-derived recombinant human LILRB2/CD85d/ILT4 Gly24-His458 Accession # ACT64556 | |
100 μg | |
Primary | |
Detects human LILRB2/CD85d/ILT4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) ILT1, rhILT2, rhILT3, rhILT6, rhILT7, rhILT11, rhLIR6 or rhLIR8 is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. |
Flow Cytometry | |
287219 | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CD85 antigen-like family member D, CD85d, CD85d antigen, Ig-like transcript 4, ILT4, ILT-4, ILT4CD85d, Immunoglobulin-like transcript 4, Leukocyte immunoglobulin-like receptor 2, leukocyte immunoglobulin-like receptor subfamily B member 2, leukocyte immunoglobulin-like receptor subfamily B member 2 soluble isoform, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR2, LIR2CD85D, LIR-2subfamily A (with TM domain), member 6, member 2, MIR10, MIR10LILRA6, Monocyte/macrophage immunoglobulin-like receptor 10 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
10288 | |
Human | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction