Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human LILRB2/CD85d/ILT4 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems BAF2078
This item is not returnable.
View return policy
Description
ILT4/CD85d/LILRB2 Polyclonal specifically detects ILT4/CD85d/LILRB2 in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
ILT4/CD85d/LILRB2 | |
Polyclonal | |
Biotin | |
AAC51882 | |
LILRB2 | |
Mouse myeloma cell line NS0-derived recombinant human LILRB2/CD85d/ILT4 Gly24-His458 Accession # AAC51882 | |
50 μg | |
Primary | |
Detects human LILRB2/CD85d/ILT4 in Western blots. In Western blots, approximately 30% cross-reactivity with recombinant human (rh) ILT2 is observed and less than 5% cross-reactivity with rhILT5 is observed. | |
Human | |
IgG |
Western Blot, Flow Cytometry | |
LYOPH | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells | |
CD85 antigen-like family member D, CD85d, CD85d antigen, Ig-like transcript 4, ILT4, ILT-4, ILT4CD85d, Immunoglobulin-like transcript 4, Leukocyte immunoglobulin-like receptor 2, leukocyte immunoglobulin-like receptor subfamily B member 2, leukocyte immunoglobulin-like receptor subfamily B member 2 soluble isoform, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR2, LIR2CD85D, LIR-2subfamily A (with TM domain), member 6, member 2, MIR10, MIR10LILRA6, Monocyte/macrophage immunoglobulin-like receptor 10 | |
Goat | |
Affinity Purified | |
RUO | |
10288 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction